Skip to Content
Merck
IdMOC® tissue culture plate 16 chambers, size 96 wells, pkg of 5 ea

IdMOC® tissue culture plate

IdMOC® tissue culture plate 16 chambers, size 96 wells, pkg of 5 ea
Synonym(s):
Integrated Descrete Multiple Organ Culture Plates

Products

Product Category
Color
Manufacturer
Binding Type
Wells
Working Volume (ml)
Color
Plate Material
Sterility