Skip to Content
Merck
Holmium(III) trifluoromethanesulfonate 98%

Holmium(III) trifluoromethanesulfonate

Holmium(III) trifluoromethanesulfonate 98%
Synonym(s):
Holmium(III) triflate, Trifluoromethanesulfonic acid holmium(III) salt
Linear Formula:
(CF3SO3)3Ho
CAS Number:
Molecular Weight:
612.14

Products

Brand
Formula Weight
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing