Skip to Content
Merck
Geranyl caproate mixture of isomers, natural (US), ≥95%, FG

Geranyl caproate

Geranyl caproate mixture of isomers, natural (US), ≥95%, FG
Synonym(s):
(E)-3,7-Dimethylocta-2,6-dienyl hexanoate, Geranyl hexanoate
Empirical Formula (Hill Notation):
C16H28O2
CAS Number:
Molecular Weight:
252.39
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing