Skip to Content
Merck
Fmoc-Phe-OPfp Novabiochem®

Fmoc-Phe-OPfp

Fmoc-Phe-OPfp Novabiochem®
Synonym(s):
Fmoc-Phe-OPfp
Empirical Formula (Hill Notation):
C30H20F5NO4
CAS Number:
Molecular Weight:
553.48

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing