Fenofibrate impurity G
Synonym(s):
2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoic acid 1,1-dimethyl-2-(1-methylethoxy)-2-oxoethyl ester
Empirical Formula (Hill Notation):
C24H27ClO6
CAS Number:
Molecular Weight:
446.92
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing