Skip to Content
Merck
D-Pantethine analytical standard

D-Pantethine

D-Pantethine analytical standard
Synonym(s):
Bis(N-pantothenylamidoethyl) disulfide, LBF factor, Pantethein oxidized form
Empirical Formula (Hill Notation):
C22H42N4O8S2
CAS Number:
Molecular Weight:
554.72
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing