Dicyclanil
Synonym(s):
4,6-Diamino-2-(cyclopropylamino)-5-pyrimidinecarbonitrile
Empirical Formula (Hill Notation):
C8H10N6
CAS Number:
Molecular Weight:
190.21
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing