Skip to Content
Merck
δ-Decalactone natural, ≥98%, FCC, FG

δ-Decalactone

δ-Decalactone natural, ≥98%, FCC, FG
Synonym(s):
(±)-5-Decanolide, δ-Decanolactone, (±)-δ-Pentyl-δ-valerolactone, (±)-6-Pentyltetrahydro-2H-pyran-2-one, 5-Hydroxydecanoic acid δ-lactone
Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing