![D-Camphor Impurity I Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/365/164/8ed35c15-3247-4ee9-ae4b-96b685743975/640/8ed35c15-3247-4ee9-ae4b-96b685743975.png)
D-Camphor Impurity I
![D-Camphor Impurity I Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/365/164/8ed35c15-3247-4ee9-ae4b-96b685743975/640/8ed35c15-3247-4ee9-ae4b-96b685743975.png)
Synonym(s):
Isoborneol
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing