Skip to Content
Merck
Dapagliflozin propanediol monohydrate ≥98% (HPLC)

Dapagliflozin propanediol monohydrate

Dapagliflozin propanediol monohydrate ≥98% (HPLC)
Synonym(s):
(1S)-1,5-Anhydro-1-C-[4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-D-glucitol (S)-propane-1,2-diol (1:1) monohydrate, (2S,3R,4R,5S,6R)-2-(3-(4-Ethoxybenzyl)-4-chlorophenyl)-6-hydroxymethyl-tetrahydro-2H-pyran-3,4,5-triol propanediol monohydrate, BMS 512148 propanediol hydrate, BMS-512148 propanediol hydrate, Dapagliflozin (S)-propylene glycol monohydrate, Dapagliflozin propanediol hydrate
Empirical Formula (Hill Notation):
C21H25ClO6·C3H8O2·H2O
CAS Number:
Molecular Weight:
502.98

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing