Skip to Content
Merck
Candicidin ≥50% (HPLC)

Candicidin

Candicidin ≥50% (HPLC)
Synonym(s):
Candicidin
Empirical Formula (Hill Notation):
C59H84N2O18
CAS Number:
Molecular Weight:
1109.30

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing