![Calmodulin-Agarose saline suspension](/deepweb/assets/sigmaaldrich/product/images/373/840/29cb577b-3ffe-4767-ac63-c0350c91ec48/640/29cb577b-3ffe-4767-ac63-c0350c91ec48.jpg)
Calmodulin-Agarose
![Calmodulin-Agarose saline suspension](/deepweb/assets/sigmaaldrich/product/images/373/840/29cb577b-3ffe-4767-ac63-c0350c91ec48/640/29cb577b-3ffe-4767-ac63-c0350c91ec48.jpg)
Synonym(s):
Calmodulin−Agarose, Phosphodiesterase 3′,5′-Cyclic Nucleotide Activator-Agarose from bovine testes
Products
Brand
Biological Source
Physical Form
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing