![Boldine for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/283/906/a312c274-9c50-4603-83c4-56d042b08d17/640/a312c274-9c50-4603-83c4-56d042b08d17.png)
Boldine for system suitability
![Boldine for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/283/906/a312c274-9c50-4603-83c4-56d042b08d17/640/a312c274-9c50-4603-83c4-56d042b08d17.png)
Synonym(s):
Boldine, 2,9-Dihydroxy-1,10-dimethoxyaporphine
Empirical Formula (Hill Notation):
C19H21NO4
CAS Number:
Molecular Weight:
327.37
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing