
Anti-GATA1 antibody produced in rabbit

Synonym(s):
Eryf1, Erythroid transcription factor, GATA-1, GATA-binding factor 1, GF-1, Anti-ERYF1, Anti-GATA binding protein 1 (globin transcription factor 1), Anti-GF1, Anti-NFE1
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing