
Allopurinol Related Compound D

Synonym(s):
Ethyl 3-aminopyrazole-4-carboxylate, 3-Amino-4-carbethoxypyrazole
Empirical Formula (Hill Notation):
C6H9N3O2
CAS Number:
Molecular Weight:
155.15
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2