
(Acetylacetonato)(1,5-cyclooctadiene)iridium(I)

Synonym(s):
Ir(acac)(COD)
Empirical Formula (Hill Notation):
C13H19IrO2
CAS Number:
Molecular Weight:
399.51
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing