![5-Chloro-3-pyridinamine AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/439/586/17cc5874-4cbc-488c-8e8c-34152a9a2650/640/17cc5874-4cbc-488c-8e8c-34152a9a2650.png)
5-Chloro-3-pyridinamine
![5-Chloro-3-pyridinamine AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/439/586/17cc5874-4cbc-488c-8e8c-34152a9a2650/640/17cc5874-4cbc-488c-8e8c-34152a9a2650.png)
Empirical Formula (Hill Notation):
C5H5ClN2
CAS Number:
Molecular Weight:
128.56
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing