
4-Chlorophenyl chloroformate

Linear Formula:
ClCO2C6H4Cl
CAS Number:
Molecular Weight:
191.01
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing