Skip to Content
Merck
vibrant-m

4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one solution

vibrant-m
Synonym(s):
Caramel furanone, Sotolon, Sugar lactone
Empirical Formula (Hill Notation):
C6H8O3
CAS Number:
Molecular Weight:
128.13

Products

Product Category
Brand
Biological Source
Formula Weight
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1