Skip to Content
Merck
3,5-Diiodo-L-tyrosine dihydrate crystalline

3,5-Diiodo-L-tyrosine dihydrate

3,5-Diiodo-L-tyrosine dihydrate crystalline
Empirical Formula (Hill Notation):
C9H9I2NO3 · 2H2O
CAS Number:
Molecular Weight:
469.01
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Optical Isomer
Amino Acid
Technique
Showing 1-1 of 1