Skip to Content
Merck
2-Ethylhexyl salicylate analytical standard

2-Ethylhexyl salicylate

2-Ethylhexyl salicylate analytical standard
Synonym(s):
Octisalate, Octyl salicylate
Linear Formula:
(HO)C6H4CO2CH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
250.33
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing