Skip to Content
Merck
[10]-Gingerol analytical standard

10-Gingerol

[10]-Gingerol analytical standard
Synonym(s):
[10]-Gingerol, (S)- 5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-tetradecanone, 10-Gingerol
Empirical Formula (Hill Notation):
C21H34O4
CAS Number:
Molecular Weight:
350.49

Products

Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing