Product Name
4-(Azidomethyl)benzonitrile solution ~0.5 M in tert-butyl methyl ether, ≥97.0% (HPLC)
Product Number
745316
Product Brand
aldrich
CAS Number
84466-87-5
Molecular Weight
158.16
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Dark Yellow and Colorless to Dark Green-Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (HPLC AREA %) | ≥ 97.0 % |
WATER | ≤ 2.0 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
REMARKS | CONCENTRATION ~ 0.5M in TBME |