Product Name
Acetylthiocholine iodide ≥98% (TLC), powder or crystals
Product Number
A5751
Product Brand
SIGMA
CAS Number
1866-15-5
Molecular Weight
289.18
Test | Specification |
---|---|
APPEARANCE (COLOR) | White |
APPEARANCE (FORM) | Powder to Crystals |
ARGENTOMETR. TITRATION | IODIDE 42.8 - 45.0 % |
PURITY (TLC AREA %) | ≥ 98 % |
SOLUBILITY (COLOR) | Colorless |
SOLUBILITY (TURBIDITY) | Clear |
SOLUBILITY (METHOD) | 100MG/ML IN WATER |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
SUITABILITY ASSAY | SUITABLE AS SUBSTRATE FOR ACETYLCHOLINESTERASE |
RECOMMENDED RETEST PERIOD | 24 MONTHS |