Product Name
Victoria Blue B suitable for microscopy
Product Number
94890
Product Brand
SIGMA
CAS Number
2580-56-5
Molecular Weight
506.08
Test | Specification |
---|---|
APPEARANCE (COLOR) | Blue to Very Dark Blue and Purple to Very Dark Purple and Black |
APPEARANCE (FORM) | Powder |
THIN LAYER CHROMATOGR. | CORRESPONDS TO STANDARD |
SOLUBILITY (COLOR) | Blue to Very Dark Blue |
SOLUBILITY (TURBIDITY) | Report Result |
SOLUBILITY (METHOD) | 0.01G IN 10ML METHANOL |
WAVELENGTH (1) (UV) | 594.0 - 609.0 nm |
EXTINCTION (1%/1CM) (1) (UV) | ≥ 1200 |
SOLVENT (UV) | Ethanol |