Pressure Conversion Table
Pressure Unit Conversion Table & Chart
Use this pressure conversion table chart to convert pressure units. To use the chart, find the unit of pressure you want to convert in the left hand column and multiply it by the value in the corresponding pressure unit column.
The handy pressure converter table is used for converting pressure units fast and easily in the lab or in the classrom. All you need is a calculator.
This chart is available in downloadable format for printing and republishing with a reference back to this page.
kPa | bar | atm | Torr | psi | mmHg | |
---|---|---|---|---|---|---|
kPa | 1 | 0.01 | 0.00987 | 7.5 | 0.146 | 7.5 |
cm3 | 100 | 1 | 0.987 | 750.06 | 14.504 | 750.06 |
m3 | 101.325 | 1.013 | 1 | 760 | 14.696 | 760 |
in3 | 0.133 | 0.00133 | 0.00132 | 1 | 0.0193 | 1 |
ft3 | 6.895 | 0.069 | 0.068 | 51.715 | 1 | 51.715 |
yd3 | 0.133 | 0.0013 | 0.0013 | 1 | 0.0193 | 1 |
Pressure Conversion Multiplication Chart
This expanded pressure conversion chart lists all conversion formulas for pressure units. This chart is available to download in PDF format.
Convert From | To | Multiply by | Convert From | To | Multiply by |
---|---|---|---|---|---|
bar | psi | 14.5 | micron (μ) | psi | 0.00019337 |
bar | in. of H20 | 401.47 | micron (μ) | in. of H2O | 0.0053525 |
bar | in. of Hg | 29.53 | micron (μ) | in. of Hg | 0.0003937 |
bar | kPa | 100 | micron (μ) | kPa | 0.0013332 |
bar | mbar | 1000 | micron (μ) | mbar | 0.013332 |
bar | cm of H20 | 1019.73 | micron (μ) | cm of H2O | 0.013595 |
bar | mm of Hg | 750.06 | micron (μ) | mm of Hg | 0.001 |
in. of H20 | psi | 0.036127 | mm of Hg | psi | 0.019337 |
in. of H20 | in. of Hg | 0.073554 | mm of Hg | in. of H2O | 0.53525 |
in. of H20 | kPa | 0.2491 | mm of Hg | in. of Hg | 0.03937 |
in. of H20 | mbar | 2.491 | mm of Hg | kPa | 0.13332 |
in. of H20 | cm of H20 | 2.54 | mm of Hg | mbar | 1.3332 |
in. of H20 | mm of Hg | 1.8683 | mm of Hg | cm of H2O | 1.3595 |
in. of Hg | psi | 0.4912 | psi | in. of H2O | 27.68 |
in. of Hg | in. of H20 | 13.596 | psi | in. of Hg | 2.036 |
in. of Hg | kPa | 3.3864 | psi | kPa | 6.8947 |
in. of Hg | mbar | 33.864 | psi | mbar | 68.947 |
in. of Hg | cm of H20 | 34.532 | psi | cm of H2O | 70.308 |
in. of Hg | mm of Hg | 25.4 | psi | mm of Hg | 51.715 |
kPa | psi | 0.14504 | torr | psi | 0.019337 |
kPa | in. of H20 | 4.0147 | torr | in. of H2O | 0.53525 |
kPa | in. of Hg | 0.2953 | torr | in. of Hg | 0.03937 |
kPa | mbar | 10 | torr | kPa | 0.13332 |
kPa | cm of H20 | 10.1973 | torr | mbar | 1.3332 |
kPa | mm of Hg | 7.5006 | torr | cm of H2O | 1.3595 |
mbar | psi | 0.0145 | torr | mm of Hg | 1 |
mbar | in. of H20 | 0.40147 | |||
mbar | in. of Hg | 0.02953 | |||
mbar | kPa | 0.1 | |||
mbar | cm of H20 | 1.01973 | |||
mbar | mm of Hg | 0.75006 |