Merck
HomePhotometry & ReflectometryDetermination of Ammonium in Mineral Fertilizer
Photometric Determination of Ammonium in Mineral Fertilizer Using The Indophenol Blue Method Following an Acid Extraction
Abstract
This protocol describes the use of indophenol blue to measure ammonium in mineral fertilizers using a photometric method after an acid extraction. The approach allows precise measurement of ammonium, a critical component in nitrogenous fertilizers, which is crucial for assessing fertilizer quality and reducing their environmental impact. The process utilizes Spectroquant® test kits and spectrophotometers to ensure accurate analysis.
Section Overview:
three models of Spectroquant® spectrophotometers: Prove 100 plus, Prove 300 plus, and Prove 600 plus. Each device has a sleek, modern design with a rectangular body. The largest model in the foreground, Prove 600 plus, is green with a black top and features a touchscreen interface displaying measurement data, as well as a slot for sample insertion. The top right corner of the device has a yellow "M" logo and "600+" printed in bold yellow on the front corner. Two smaller models, Prove 100 plus and Prove 300 plus, are placed in the background. Prove 100 plus is light green with a black top, while Prove 300 plus is yellow with a black top. Both also feature touchscreens and the "M" logo.
Spectroquant® spectrophotometers - Prove 100 plus, Prove 300 plus, & Prove 600 plus
Introduction
Agriculture has been mankind’s earliest occupation and the birth and proliferation of human civilizations was possible only because of consistent evolution of agricultural methods. Improving crop yield has always been the constant stride to feed increasing populations, and in the modern age, mineral fertilizers are considerably depended on for this quest. Mineral fertilizers, with their defined concentrations of plant nutrients like nitrogen and potassium, augment healthy crop growth even on soils that are poor providers of certain mineral nutrients. Ammonium is the basic building block of commercial nitrogenous fertilizers1. It is important to analyze mineral fertilizers and soil to study accurate concentrations of specific nutrients, as their field use not only impacts crop growth but also soil health. Excessive use of nitrogenous fertilizers has shown detrimental effects to the environment, and human health2. Excess ammonium has also been reported to be toxic in higher plants3.
This application note details a photometric ammonium test method for mineral fertilizers, using the indophenol blue method after acid extraction.
Reagents, Instruments and Materials
Ammonium Test Kits/Reagents
For the measurement, one of the following Spectroquant® test kits is necessary:
  • Spectroquant® Ammonium Cell Test (1.14544)*
  • Spectroquant® Ammonium Cell Test (1.14559)
  • Spectroquant® Ammonium Test (1.14752)
  • Spectroquant® Ammonium Test (1.00683)
*not compatible with Move 100
Instrument(s) & Devices
For the measurement, one of the following Spectroquant® photometers is necessary:
  • Spectroquant® VIS Spectrophotometer Prove 100 Plus (1.73026)
  • Spectroquant® UV/VIS Spectrophotometer Prove 300 Plus (1.73027)
  • Spectroquant® UV/VIS Spectrophotometer Prove 600 Plus (1.73028)
  • Spectroquant® Colorimeter Move 100 (1.73632)
Also, legacy systems
  • Spectroquant® Spectrophotometer Prove 100/300/600
  • Spectroquant® Photometer NOVA 30/60/60A
Software for Data transfer
  • Optional Spectroquant® Prove Connect to LIMS software package (Y.11086) to transfer your data into an existing LIMS system.
Instrument Accessories
  • Rectangular cells 10 mm (1.14946) and/or
  • Rectangular cells 20 mm (1.14947) and/or
  • Rectangular cells 50 mm (1.14944)
Other Reagents and Accessories
  • Hydrochloric acid 1 mol/L (1.09057)
  • Water for analysis (1.16754)
  • Folded filter
Experimental Procedure
Sample Preparation
  • In a mill grind down about 30 g of the sample to a fine powder and homogenize.
  • Bring 0.5 g of the powdered material to boil with 25 mL water for analysis and 2.5 mL hydrochloric acid 1 mol/L.
  • After cooling, filter the mixture through a folded filter, wash the filter 3 times with boiling water for analysis, and collect the filtrate and the washings in a 500 mL volumetric flask.
  • Fill up to volume with water for analysis and mix.
  • Finally put 10 mL of this solution into a 1000 mL volumetric flask, fill up to volume, and mix.
Analysis
Determine with one of the above-mentioned test kits.
Calculation
Ammonium content in mg/kg NH4+ = analysis value in mg/L NH4+ x 10
See more applications for photometry at Protocols and Application Notes
Related Products
Lo sentimos, se ha producido un error inesperado
Network error: Failed to fetch
References
1.
Fertilizer material. [Internet]. Hawaii.edu.[cited 07 Aug 2024]. Available from: https://www.ctahr.hawaii.edu/mauisoil/c_material.aspx
2.
Ahmed M, Rauf M, Mukhtar Z, Saeed NA. 2017. Excessive use of nitrogenous fertilizers: an unawareness causing serious threats to environment and human health. Environ Sci Pollut Res. 24(35):26983-26987. https://doi.org/10.1007/s11356-017-0589-7
3.
Britto DT, Kronzucker HJ. 2002. NH4+ toxicity in higher plants: a critical review. Journal of Plant Physiology. 159(6):567-584. https://doi.org/10.1078/0176-1617-0774