Merck
HPLC Analysis of Carnitine Enantiomers on Astec® CHIROBIOTIC® TAG
HPLC Analysis of Carnitine Enantiomers on Astec® CHIROBIOTIC® TAG application for HPLC
Materiales
Analytical column
Referencia del producto
Descripción
Precios
Astec® Chirobiotic® TAG Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
standard
Referencia del producto
Descripción
Precios
(±)-Carnitine hydrochloride
≥98% (TLC)
CONDITIONS
column
CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 μm particles (14024AST)
mobile phase
[A] 5 mM ammonium acetate, pH 6; [B] methanol; (25:75, A:B)
flow rate
0.6 mL/min
column temp.
35 °C
detector
ESI(+)-MS (m/z = 162)
injection
5 μL
sample
1 μg/mL in mobile phase