Merck
C11orf95-RELA fusions drive oncogenic NF-κB signalling in ependymoma.
Nature (2014-02-21)
Matthew Parker, Kumarasamypet M Mohankumar, Chandanamali Punchihewa, Ricardo Weinlich, James D Dalton, Yongjin Li, Ryan Lee, Ruth G Tatevossian, Timothy N Phoenix, Radhika Thiruvenkatam, Elsie White, Bo Tang, Wilda Orisme, Kirti Gupta, Michael Rusch, Xiang Chen, Yuxin Li, Panduka Nagahawhatte, Erin Hedlund, David Finkelstein, Gang Wu, Sheila Shurtleff, John Easton, Kristy Boggs, Donald Yergeau, Bhavin Vadodaria, Heather L Mulder, Jared Becksfort, Jared Becksford, Pankaj Gupta, Robert Huether, Jing Ma, Guangchun Song, Amar Gajjar, Thomas Merchant, Frederick Boop, Amy A Smith, Li Ding, Charles Lu, Kerri Ochoa, David Zhao, Robert S Fulton, Lucinda L Fulton, Elaine R Mardis, Richard K Wilson, James R Downing, Douglas R Green, Jinghui Zhang, David W Ellison, Richard J Gilbertson
RESUMEN
Members of the nuclear factor-κB (NF-κB) family of transcriptional regulators are central mediators of the cellular inflammatory response. Although constitutive NF-κB signalling is present in most human tumours, mutations in pathway members are rare, complicating efforts to understand and block aberrant NF-κB activity in cancer. Here we show that more than two-thirds of supratentorial ependymomas contain oncogenic fusions between RELA, the principal effector of canonical NF-κB signalling, and an uncharacterized gene, C11orf95. In each case, C11orf95-RELA fusions resulted from chromothripsis involving chromosome 11q13.1. C11orf95-RELA fusion proteins translocated spontaneously to the nucleus to activate NF-κB target genes, and rapidly transformed neural stem cells--the cell of origin of ependymoma--to form these tumours in mice. Our data identify a highly recurrent genetic alteration of RELA in human cancer, and the C11orf95-RELA fusion protein as a potential therapeutic target in supratentorial ependymoma.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-L1CAM antibody, Mouse monoclonal, clone UJ127.11, purified from hybridoma cell culture