Merck
  • Three-dimensional electron microscopy reconstruction and cysteine-mediated crosslinking provide a model of the type III secretion system needle tip complex.
Three-dimensional electron microscopy reconstruction and cysteine-mediated crosslinking provide a model of the type III secretion system needle tip complex.
Molecular microbiology (2014-10-30)
Martin Cheung, Da-Kang Shen, Fumiaki Makino, Takayuki Kato, A Dorothea Roehrich, Isabel Martinez-Argudo, Matthew L Walker, Isabel Murillo, Xia Liu, Maria Pain, James Brown, Gordon Frazer, Judith Mantell, Petros Mina, Thomas Todd, Richard B Sessions, Keiichi Namba, Ariel J Blocker
RESUMEN
Type III secretion systems are found in many Gram-negative bacteria. They are activated by contact with eukaryotic cells and inject virulence proteins inside them. Host cell detection requires a protein complex located at the tip of the device's external injection needle. The Shigella tip complex (TC) is composed of IpaD, a hydrophilic protein, and IpaB, a hydrophobic protein, which later forms part of the injection pore in the host membrane. Here we used labelling and crosslinking methods to show that TCs from a ΔipaB strain contain five IpaD subunits while the TCs from wild-type can also contain one IpaB and four IpaD subunits. Electron microscopy followed by single particle and helical image analysis was used to reconstruct three-dimensional images of TCs at ∼ 20 Å resolution. Docking of an IpaD crystal structure, constrained by the crosslinks observed, reveals that TC organisation is different from that of all previously proposed models. Our findings suggest new mechanisms for TC assembly and function. The TC is the only site within these secretion systems targeted by disease-protecting antibodies. By suggesting how these act, our work will allow improvement of prophylactic and therapeutic strategies.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment−R-Phycoerythrin antibody produced in goat, affinity isolated antibody, buffered aqueous solution