Merck
  • IGF1/insulin receptor kinase inhibition by BMS-536924 is better tolerated than alloxan-induced hypoinsulinemia and more effective than metformin in the treatment of experimental insulin-responsive breast cancer.
IGF1/insulin receptor kinase inhibition by BMS-536924 is better tolerated than alloxan-induced hypoinsulinemia and more effective than metformin in the treatment of experimental insulin-responsive breast cancer.
Endocrine-related cancer (2011-09-29)
Carly Jade Dool, Haider Mashhedi, Mahvash Zakikhani, Stéphanie David, Yunhua Zhao, Elena Birman, Joan M Carboni, Marco Gottardis, Marie-José Blouin, Michael Pollak
RESUMEN
Epidemiologic and experimental evidence suggest that a subset of breast cancer is insulin responsive, but it is unclear whether safe and effective therapies that target the insulin receptor (IR), which is homologous to oncogenes of the tyrosine kinase class, can be developed. We demonstrate that both pharmacologic inhibition of IR family tyrosine kinase activity and insulin deficiency have anti-neoplastic activity in a model of insulin-responsive breast cancer. Unexpectedly, in contrast to insulin deficiency, pharmacologic IR family inhibition does not lead to significant hyperglycemia and is well tolerated. We show that pharmacokinetic factors explain the tolerability of receptor inhibition relative to insulin deficiency, as the small molecule receptor kinase inhibitor BMS-536924 does not accumulate in muscle at levels sufficient to block insulin-stimulated glucose uptake. Metformin, which lowers insulin levels only in settings of hyperinsulinemia, had minimal activity in this normoinsulinemic model. These findings highlight the importance of tissue-specific drug accumulation as a determinant of efficacy and toxicity of tyrosine kinase inhibitors and suggest that therapeutic targeting of the IR family for cancer treatment is practical.