Merck
  • Combined Bone Mesenchymal Stem Cell and Olfactory Ensheathing Cell Transplantation Promotes Neural Repair Associated With CNTF Expression in Traumatic Brain-Injured Rats.
Combined Bone Mesenchymal Stem Cell and Olfactory Ensheathing Cell Transplantation Promotes Neural Repair Associated With CNTF Expression in Traumatic Brain-Injured Rats.
Cell transplantation (2014-03-13)
Xue-Mei Fu, Su-Juan Liu, Qi-Qin Dan, Yan-Ping Wang, Na Lin, Long-Yun Lv, Yu Zou, Su Liu, Xue Zhou, Ting-Hua Wang
RESUMEN
This study examined the role of bone mesenchymal stem cell (BMSC) and olfactory ensheathing cell (OEC) cografting on neural function and underlying molecular mechanisms in acute stage of traumatic brain injury (TBI) rats. Eighty Sprague-Dawley (SD) female rats were randomly divided into five groups (n = 16 per category): sham operated group (Sham), weight-drop-induced TBI group (TBI), BMSC transplantation group (BMSC), OEC transplantation group (OEC), and cotransplantation group (CO). Eight rats were randomly selected from each group for behavioral and morphological assessment. Another category (n = 8 rats) was employed in the genetic expression detection. BMSCs were isolated from GFP mice and identified by CD44 antibody. OECs were isolated from the SD rats, identified by P75 antibody and labeled by Hoechst 33342. They were then transplanted into the surrounding tissue of the epicenter of TBI rats. The result of neurological severity scores revealed that BMSC or OEC transplantation alone and BMSC and OEC cografting significantly ameliorated the neurological deficits of TBI rats. Quantitative immunohistochemical analysis showed that graft-recipient animals possessed dramatically more neurons and regenerated axons and smaller amounts of astrocytes than controls 14 days posttransplantation (p < 0.05). However, the expressional level of ciliary neurotrophic factor significantly decreased in the cografting group as determined by RT-PCR (p < 0.05), and the Janus kinase/signal transducer and activator of transcription pathway was significantly activated at 7 days after cell transplantation (p < 0.05). This study is the first to report the role of cotransplantation of BMSCs and OECs in the therapy of TBI and explore its potential molecular mechanisms, therefore providing the important morphological and molecular biological evidence for the clinical application of BMSC and/or OEC transplantation in TBI.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sacarosa, for molecular biology, ≥99.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Azul de tripano solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioXtra
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dietilpirocarbonato, 96% (NT)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, BioUltra, for molecular biology, ≥99.5% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Forskolina, from Coleus forskohlii, ≥98% (HPLC), powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Forskolina, For use in molecular biology applications
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), Grade II, suitable for plant cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dietilpirocarbonato, 96% (NT)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, Grade I, ≥99% (GC), suitable for plant cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dietilpirocarbonato, 96% (NT)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, meets USP testing specifications
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Chloral hydrate, ≥99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ACS reagent
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, puriss., meets analytical specification of Ph. Eur., BP, NF
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
(±)-CPP, solid
En este momento no podemos mostrarle ni los precios ni la disponibilidad