Merck
Sex and estrogen affect the distribution of urocortin-1 immunoreactivity in brainstem autonomic nuclei of the rat.
Brain research bulletin (2015-07-07)
John Ciriello
RESUMEN
Urocortin-1 (UCN-1), a neuropeptide closely related to the hypothalamic hormone corticotropin-releasing factor, has been associated with stress, feeding behaviors, cardiovascular control, and to exhibit functional gender differences. This study was done to investigate whether estrogen (E; 17β-estradiol) treatment (9 weeks) altered UCN-1 immunoreactivity in brainstem autonomic nuclei in female Wistar rats. Experiments were done in age matched adult males (controls), females (intact), and ovariectomized (OVX) only and OVX+E (30pg/ml plasma) treated females. All animals received intracerebroventricular injections of colchicine and were then perfused transcardially with Zamboni's fixative. Coronal brainstem sections (40μm) were cut and processed immunohistochemically for UCN-1. In males, moderate UCN-1 fiber labeling was found in the nucleus of the solitary tract (NTS) and throughout the rostral ventral lateral medulla (RVLM). Additionally, a few UCN-1 immunoreactive neurons were observed in hypoglossal nucleus (XII), facial nucleus (FN) and nucleus ambiguus (Amb). In intact females and OVX+E females, fewer UCN-1 labeled fibers were found within NTS compared to males. In contrast, the RVLM was more densely innervated in the female cases. Furthermore, in both intact and OVX+E females UCN-1 labeled neurons were found not only within Amb, FN and XII, but also within NTS, RVLM and nucleus raphé pallidus (RP). In OVX only animals, moderate to dense UCN-1 fiber labeling was observed in the NTS complex and throughout RVLM compared to males and the other female groups. However, in contrast to all other groups, UCN-1 labeled neurons were found in greater number within Amb, FN, NTS, dorsal motor nucleus of the vagus, XII, RVLM, magnocellular reticular nucleus and RP. These data not only suggest that sex differences exist in the distribution of UCN-1 within brainstem autonomic areas, but that circulating level of E may play an important role with regards to the function of these UCN-1 neurons during stress responses.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Peróxido de hidrógeno solution, 30 % (w/w) in H2O, contains stabilizer
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, for molecular biology, ≥99.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Colesterol, Sigma Grade, ≥99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioXtra
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, BioUltra, for molecular biology, ≥99.5% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Colchicina, ≥95% (HPLC), powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Colesterol, powder, BioReagent, suitable for cell culture, ≥99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Colesterol, from sheep wool, ≥92.5% (GC), powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), Grade II, suitable for plant cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Colchicina, BioReagent, suitable for plant cell culture, ≥95% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, Grade I, ≥99% (GC), suitable for plant cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
SAFC
Colesterol, derivado vegetal, SyntheChol®
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, meets USP testing specifications
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Peróxido de hidrógeno solution, 34.5-36.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ACS reagent
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, puriss., meets analytical specification of Ph. Eur., BP, NF
En este momento no podemos mostrarle ni los precios ni la disponibilidad
SAFC
Colesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ampicillin, meets USP testing specifications
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Peróxido de hidrógeno solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
En este momento no podemos mostrarle ni los precios ni la disponibilidad