Merck
  • Prostacyclin receptor (PTGIR) in the porcine endometrium: Regulation of expression and role in luminal epithelial and stromal cells.
Prostacyclin receptor (PTGIR) in the porcine endometrium: Regulation of expression and role in luminal epithelial and stromal cells.
Theriogenology (2015-07-04)
Agnieszka Blitek, Magdalena Szymanska, Ewa Morawska-Pucinska, Izabela Malysz-Cymborska, Aneta Andronowska
RESUMEN
The prostacyclin (PGI2) signaling pathway plays an important role during early pregnancy in rodents and ruminants. Abundant concentrations of PGI2 were also found in the endometrium and uterine lumen of gilts during the period of implantation. The present study was designed to examine (1) the expression of PGI2 receptor (PTGIR) messenger RNA (mRNA) and protein in the endometrium of cyclic and early-pregnant gilts; (2) possible regulation of endometrial PTGIR gene expression by conceptus products, estradiol and cytokines; (3) the effect of iloprost (a PGI2 analogue) on cAMP formation and the expression of fibroblast growth factor-2 (FGF-2) and vascular endothelial growth factor (VEGF; isoform 164) mRNA in luminal epithelial (LE) and stromal (ST) cells. Increased PTGIR mRNA expression in the endometrium was detected on Days 11 to 12 and 18 to 20 of pregnancy compared with respective days of the estrous cycle (P < 0.05). Moreover, greater PTGIR protein level was observed in pregnant than in cyclic gilts on Days 11 to 12 after estrus (P < 0.05). Gilts with unilateral pregnancy revealed abundant PTGIR expression in the endometrium collected from the gravid uterine horn of Day-11 pregnant gilts compared with the respective horn of cyclic animals (P < 0.01). Both LE and ST cells of the endometrium possess PTGIR protein. Moreover, IL1β, IFNγ, and conceptus-exposed medium, but not estradiol, stimulated PTGIR mRNA expression in LE and ST cells in vitro. Activation of PTGIR by incubation of LE and ST cells with iloprost resulted in greater cAMP generation (P < 0.01). Moreover, iloprost increased FGF-2 and VEGF164 mRNA expression in ST (P < 0.05), but not LE cells. In conclusion, this study revealed increased expression of PTGIR in the porcine endometrium during the peri-implantation period and reported a possible regulation of PTGIR abundance by conceptus-derived factors. Moreover, besides its important role in vascular system, PGI2 may promote the expression of proangiogenic genes in the uterine stroma.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
DAPI, for nucleic acid staining
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Cloruro de sodio, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Cloruro de sodio, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Fenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
SAFC
Sodium chloride solution, 5 M
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Cloruro de sodio, 99.999% trace metals basis
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Fenol, ≥99%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Fenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Cloruro de sodio, BioUltra, for molecular biology, ≥99.5% (AT)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
N-Chlorosuccinimide, 98%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Triton X-100, laboratory grade
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Fenol, natural, 97%, FG
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
tert-Butyl acetoacetate, reagent grade, 98%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Cloruro de sodio, BioXtra, ≥99.5% (AT)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Fenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Cloruro de sodio, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Fenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Fenol solution, ≥89.0%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Cloruro de sodio, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Cloruro de sodio, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad