Merck
Structural and mechanistic roles of novel chemical ligands on the SdiA quorum-sensing transcription regulator.
mBio (2015-04-02)
Y Nguyen, Nam X Nguyen, Jamie L Rogers, Jun Liao, John B MacMillan, Youxing Jiang, Vanessa Sperandio
RESUMEN
Bacteria engage in chemical signaling, termed quorum sensing (QS), to mediate intercellular communication, mimicking multicellular organisms. The LuxR family of QS transcription factors regulates gene expression, coordinating population behavior by sensing endogenous acyl homoserine lactones (AHLs). However, some bacteria (such as Escherichia coli) do not produce AHLs. These LuxR orphans sense exogenous AHLs but also regulate transcription in the absence of AHLs. Importantly, this AHL-independent regulatory mechanism is still largely unknown. Here we present several structures of one such orphan LuxR-type protein, SdiA, from enterohemorrhagic E. coli (EHEC), in the presence and absence of AHL. SdiA is actually not in an apo state without AHL but is regulated by a previously unknown endogenous ligand, 1-octanoyl-rac-glycerol (OCL), which is ubiquitously found throughout the tree of life and serves as an energy source, signaling molecule, and substrate for membrane biogenesis. While exogenous AHL renders to SdiA higher stability and DNA binding affinity, OCL may function as a chemical chaperone placeholder that stabilizes SdiA, allowing for basal activity. Structural comparison between SdiA-AHL and SdiA-OCL complexes provides crucial mechanistic insights into the ligand regulation of AHL-dependent and -independent function of LuxR-type proteins. Importantly, in addition to its contribution to basic science, this work has implications for public health, inasmuch as the SdiA signaling system aids the deadly human pathogen EHEC to adapt to a commensal lifestyle in the gastrointestinal (GI) tract of cattle, its main reservoir. These studies open exciting and novel avenues to control shedding of this human pathogen in the environment. Quorum sensing refers to bacterial chemical signaling. The QS acyl homoserine lactone (AHL) signals are recognized by LuxR-type receptors that regulate gene transcription. However, some bacteria have orphan LuxR-type receptors and do not produce AHLs, sensing them from other bacteria. We solved three structures of the E. coli SdiA orphan, in the presence and absence of AHL. SdiA with no AHL is not in an apo state but is regulated by a previously unknown endogenous ligand, 1-octanoyl-rac-glycerol (OCL). OCL is ubiquitously found in prokaryotes and eukaryotes and is a phospholipid precursor for membrane biogenesis and a signaling molecule. While AHL renders to SdiA higher stability and DNA-binding affinity, OCL functions as a chemical chaperone placeholder, stabilizing SdiA and allowing for basal activity. Our studies provide crucial mechanistic insights into the ligand regulation of SdiA activity.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Glicerol, for molecular biology, ≥99.0%
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Cloruro de sodio, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Cloruro de sodio, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Imidazol, for molecular biology, ≥99% (titration)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
SAFC
Sodium chloride solution, 5 M
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Glicerol solution, 83.5-89.5% (T)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Glicerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Glicerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Imidazol, BioUltra, ≥99.5% (GC)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Cloruro de sodio, 99.999% trace metals basis
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Glicerol, FCC, FG
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Cloruro de sodio, BioUltra, for molecular biology, ≥99.5% (AT)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Glicerol, ≥99.5%
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Cloruro de sodio, BioXtra, ≥99.5% (AT)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Imidazol, ≥99% (titration), crystalline
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Imidazol, ACS reagent, ≥99% (titration)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
Cloruro de sodio, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
1 ea
Normalmente se envía en 1 semana. (Para pedidos fuera de Estados Unidos y Europa, calcule la entrega 1 o 2 semanas más tarde)
USD 500.00