Merck
  • Fluoxetine induces input-specific hippocampal dendritic spine remodeling along the septotemporal axis in adulthood and middle age.
Fluoxetine induces input-specific hippocampal dendritic spine remodeling along the septotemporal axis in adulthood and middle age.
Hippocampus (2015-04-09)
Kathleen McAvoy, Craig Russo, Shannen Kim, Genelle Rankin, Amar Sahay
RESUMEN
Fluoxetine, a selective serotonin-reuptake inhibitor (SSRI), is known to induce structural rearrangements and changes in synaptic transmission in hippocampal circuitry. In the adult hippocampus, structural changes include neurogenesis, dendritic, and axonal plasticity of pyramidal and dentate granule neurons, and dedifferentiation of dentate granule neurons. However, much less is known about how chronic fluoxetine affects these processes along the septotemporal axis and during the aging process. Importantly, studies documenting the effects of fluoxetine on density and distribution of spines along different dendritic segments of dentate granule neurons and CA1 pyramidal neurons along the septotemporal axis of hippocampus in adulthood and during aging are conspicuously absent. Here, we use a transgenic mouse line in which mature dentate granule neurons and CA1 pyramidal neurons are genetically labeled with green fluorescent protein (GFP) to investigate the effects of chronic fluoxetine treatment (18 mg/kg/day) on input-specific spine remodeling and mossy fiber structural plasticity in the dorsal and ventral hippocampus in adulthood and middle age. In addition, we examine levels of adult hippocampal neurogenesis, maturation state of dentate granule neurons, neuronal activity, and glutamic acid decarboxylase-67 expression in response to chronic fluoxetine in adulthood and middle age. Our studies reveal that while chronic fluoxetine fails to augment adult hippocampal neurogenesis in middle age, the middle-aged hippocampus retains high sensitivity to changes in the dentate gyrus (DG) such as dematuration, hypoactivation, and increased glutamic acid decarboxylase 67 (GAD67) expression. Interestingly, the middle-aged hippocampus shows greater sensitivity to fluoxetine-induced input-specific synaptic remodeling than the hippocampus in adulthood with the stratum-oriens of CA1 exhibiting heightened structural plasticity. The input-specific changes and circuit-level modifications in middle-age were associated with modest enhancement in contextual fear memory precision, anxiety-like behavior and antidepressant-like behavioral responses.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sacarosa, for molecular biology, ≥99.5% (GC)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
50 g
En stock
Detalles...
USD 33.90
100 g
Fecha estimada de envío 21 de marzo de 2025
USD 57.30
250 g
Fecha estimada de envío 21 de marzo de 2025
USD 124.00
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
50 g
En stock
Detalles...
USD 33.90
100 g
Fecha estimada de envío 21 de marzo de 2025
USD 57.30
250 g
Fecha estimada de envío 21 de marzo de 2025
USD 124.00
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioXtra
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
50 g
En stock
Detalles...
USD 33.90
100 g
Fecha estimada de envío 21 de marzo de 2025
USD 57.30
250 g
Fecha estimada de envío 21 de marzo de 2025
USD 124.00
Sigma-Aldrich
Sacarosa, BioUltra, for molecular biology, ≥99.5% (HPLC)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
50 g
En stock
Detalles...
USD 33.90
100 g
Fecha estimada de envío 21 de marzo de 2025
USD 57.30
250 g
Fecha estimada de envío 21 de marzo de 2025
USD 124.00
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
50 g
En stock
Detalles...
USD 33.90
100 g
Fecha estimada de envío 21 de marzo de 2025
USD 57.30
250 g
Fecha estimada de envío 21 de marzo de 2025
USD 124.00
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
50 g
En stock
Detalles...
USD 33.90
100 g
Fecha estimada de envío 21 de marzo de 2025
USD 57.30
250 g
Fecha estimada de envío 21 de marzo de 2025
USD 124.00
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), Grade II, suitable for plant cell culture
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
50 g
En stock
Detalles...
USD 33.90
100 g
Fecha estimada de envío 21 de marzo de 2025
USD 57.30
250 g
Fecha estimada de envío 21 de marzo de 2025
USD 124.00
Sigma-Aldrich
Sacarosa, Grade I, ≥99% (GC), suitable for plant cell culture
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
50 g
En stock
Detalles...
USD 33.90
100 g
Fecha estimada de envío 21 de marzo de 2025
USD 57.30
250 g
Fecha estimada de envío 21 de marzo de 2025
USD 124.00
Sigma-Aldrich
Sacarosa, meets USP testing specifications
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
50 g
En stock
Detalles...
USD 33.90
100 g
Fecha estimada de envío 21 de marzo de 2025
USD 57.30
250 g
Fecha estimada de envío 21 de marzo de 2025
USD 124.00
Sigma-Aldrich
Sacarosa, ACS reagent
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
50 g
En stock
Detalles...
USD 33.90
100 g
Fecha estimada de envío 21 de marzo de 2025
USD 57.30
250 g
Fecha estimada de envío 21 de marzo de 2025
USD 124.00
Sigma-Aldrich
Sacarosa, puriss., meets analytical specification of Ph. Eur., BP, NF
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
50 g
En stock
Detalles...
USD 33.90
100 g
Fecha estimada de envío 21 de marzo de 2025
USD 57.30
250 g
Fecha estimada de envío 21 de marzo de 2025
USD 124.00