Merck
Replacing Non-Active Video Gaming by Active Video Gaming to Prevent Excessive Weight Gain in Adolescents.
PloS one (2015-07-15)
Monique Simons, Johannes Brug, Mai J M Chinapaw, Michiel de Boer, Jaap Seidell, Emely de Vet
RESUMEN
The aim of the current study was to evaluate the effects of and adherence to an active video game promotion intervention on anthropometrics, sedentary screen time and consumption of sugar-sweetened beverages and snacks among non-active video gaming adolescents who primarily were of healthy weight. We assigned 270 gaming (i.e. ≥ 2 hours/week non-active video game time) adolescents randomly to an intervention group (n = 140) (receiving active video games and encouragement to play) or a waiting-list control group (n = 130). BMI-SDS (SDS = adjusted for mean standard deviation score), waist circumference-SDS, hip circumference and sum of skinfolds were measured at baseline, at four and ten months follow-up (primary outcomes). Sedentary screen time, physical activity, consumption of sugar-sweetened beverages and snacks, and process measures (not at baseline) were assessed with self-reports at baseline, one, four and ten months follow-up. Multi-level-intention to treat-regression analyses were conducted. The control group decreased significantly more than the intervention group on BMI-SDS (β = 0.074, 95%CI: 0.008;0.14), and sum of skinfolds (β = 3.22, 95%CI: 0.27;6.17) (overall effects). The intervention group had a significantly higher decrease in self-reported non-active video game time (β = -1.76, 95%CI: -3.20;-0.32) and total sedentary screen time (Exp (β = 0.81, 95%CI: 0.74;0.88) than the control group (overall effects). The process evaluation showed that 14% of the adolescents played the Move video games every week ≥ 1 hour/week during the whole intervention period. The active video game intervention did not result in lower values on anthropometrics in a group of 'excessive' non-active video gamers (mean ~ 14 hours/week) who primarily were of healthy weight compared to a control group throughout a ten-month-period. Even some effects in the unexpected direction were found, with the control group showing lower BMI-SDS and skin folds than the intervention group. The intervention did result in less self-reported sedentary screen time, although these results are likely biased by social desirability. Dutch Trial Register NTR3228.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sacarosa, for molecular biology, ≥99.5% (GC)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioXtra
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sacarosa, BioUltra, for molecular biology, ≥99.5% (HPLC)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), Grade II, suitable for plant cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sacarosa, Grade I, ≥99% (GC), suitable for plant cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sacarosa, meets USP testing specifications
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sacarosa, ACS reagent
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sacarosa, puriss., meets analytical specification of Ph. Eur., BP, NF
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad