Merck
  • Exfoliative cytology as a tool for monitoring pre-malignant and malignant lesions based on combined stains and morphometry techniques.
Exfoliative cytology as a tool for monitoring pre-malignant and malignant lesions based on combined stains and morphometry techniques.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology (2014-07-30)
Ignacio Gonzalez Segura, Dante Secchi, Andres Carrica, Rosario Barello, Dario Arbelo, Adriana Burgos, Mabel Brunotto, Ana M Zarate
RESUMEN
Prevention and early diagnosis have the greatest potential for public health and are the most effective method in the long-term to control oral cancer. The aim was to apply PAP staining together with AgNOR staining and morphometric analysis in oral exfoliative cytology, to determine the sensitivity and specificity of these methods in the detection of malignant changes for the purposes of both initial population monitoring and follow-up. AgNOR, Papanicolau, and morphometric tests were conducted in samples of patients with oral cancer, oral potentially malignant disorders and controls (opposite side of lesions). Specificity and sensitivity values for each stain method and the curve under ROC area were estimated. The diagnostic variables which allowed greatest accuracy in identifying malignancy relative to the healthy control were cluster (76.92%), satellite (75.64%), and total (90%). The diagnosis was seen to be associated with PAP and total AgNOR, total AgNOR and PAP, total AgNOR and satellites and clusters, and total AgNOR nuclear area/cytoplasmic area ratio. The total number of AgNOR is a reliable marker for detecting neoplastic cells; this method increases sensitivity and specificity by decreasing the likelihood of false negatives or positives, as the accuracy obtained was 90%. It is also a low-cost, non-invasive, simple methodology that can be recommended to help the early detection of oral cancer and monitoring of patients with a first diagnosis of cancer.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Alcohol etílico puro 200, for molecular biology
Sigma-Aldrich
Alcohol etílico puro, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Alcohol etílico puro, 200 proof
Sigma-Aldrich
Alcohol etílico puro, 200 proof, meets USP testing specifications
Sigma-Aldrich
Alcohol etílico puro 190, for molecular biology
Sigma-Aldrich
Etanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Alcohol etílico puro, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Etanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Alcohol etílico puro, 200 proof, anhydrous, ≥99.5%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol solution, 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Etanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Etanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
USP
Etanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Alcohol etílico puro, 190 proof, meets USP testing specifications
Supelco
Etanol, Pharmaceutical Secondary Standard; Certified Reference Material