Merck
The cytoplasmic tail of FPC antagonizes the full-length protein in the regulation of mTOR pathway.
PloS one (2014-05-24)
Shixuan Wang, Maoqing Wu, Gang Yao, Jingjing Zhang, Jing Zhou
RESUMEN
FPC (fibrocystin or polyductin) is a single transmembrane receptor-like protein, responsible for the human autosomal recessive polycystic kidney disease (ARPKD). It was recently proposed that FPC undergoes a Notch-like cleavage and subsequently the cleaved carboxy(C)-terminal fragment translocates to the nucleus. To study the functions of the isolated C-tail, we expressed the intracellular domain of human FPC (hICD) in renal epithelial cells. By 3-dimensional (3D) tubulogenesis assay, we found that in contrast to tubule-like structures formed from control cells, hICD-expressing cells exclusively formed cyst-like structures. By western blotting, we showed that the Akt/mTOR pathway, indicated by increased phosphorylation of Akt at serine 473 and S6 kinase 1 at threonine 389, was constitutively activated in hICD-expressing cells, similar to that in FPC knockdown cells and ARPKD kidneys. Moreover, application of mTOR inhibitor rapamycin reduced the size of the cyst-like structures formed by hICD-expressing cells. Application of either LY294002 or wortmannin inhibited the activation of both S6K1 and Akt. Expression of full-length FPC inhibited the activation of S6 and S6 kinase whereas co-expression of hICD with full-length FPC antagonized the inhibitory effect of full-length FPC on mTOR. Taken together, we propose that FPC modulates the PI3K/Akt/mTOR pathway and the cleaved C-tail regulates the function of the full-length protein.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
HEPES, ≥99.5% (titration)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, for molecular biology, ≥99.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
DAPI, for nucleic acid staining
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioXtra
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, BioUltra, for molecular biology, ≥99.5% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Acrilamida, suitable for electrophoresis, ≥99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Yoduro de propidio, ≥94.0% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
USP
Sacarosa, United States Pharmacopeia (USP) Reference Standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Acrilamida, for molecular biology, ≥99% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
SAFC
HEPES
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), Grade II, suitable for plant cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Acrilamida, suitable for electrophoresis, ≥99% (HPLC), powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, Grade I, ≥99% (GC), suitable for plant cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
SAFC
HEPES
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, meets USP testing specifications
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
DL-Triptófano, ≥99% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
DL-Triptófano, ≥99% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Sacarosa, Pharmaceutical Secondary Standard; Certified Reference Material
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Propidium iodide solution
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ACS reagent
En este momento no podemos mostrarle ni los precios ni la disponibilidad