Merck
Astrocytic NDRG2 is involved in glucocorticoid-mediated diabetic mechanical allodynia.
Diabetes research and clinical practice (2015-02-07)
Zhong-Fu Zuo, Yong-Hui Liao, Tan Ding, Yu-Lin Dong, Juan Qu, Jian Wang, Yan-Yan Wei, Ya-Cheng Lu, Xue-Zheng Liu, Yun-Qing Li
RESUMEN
The present study aims to test whether astrocytes contribute to glucocorticoid-mediated diabetic mechanical allodynia. Streptozotocin (STZ)-induced diabetic rats were used in our study. The intrathecal operation was performed 21 days after the onset of diabetes. Diabetic mechanical allodynia was present 28 d after the onset of diabetes, and the mechanical threshold was tested using von Frey filaments. Immunohistochemistry, including immunofluorescent histochemical staining, was performed to observe the morphology of the spinal dorsal horn (SDH). Western blot analysis was employed as a semi-quantitative assay of the expression levels of GFAP and NDRG2 associated with diabetic mechanical allodynia. Diabetic rats displayed mechanical allodynia and activated astrocytes in the SDH 28 days after the onset of diabetes. This allodynia was attenuated by intrathecal administration of the astrocyte-specific inhibitor l-α-aminoadipate. In parallel, intrathecal injection of RU486, a glucocorticoid receptor antagonist, inhibited the activation of astrocytes in the SDH, alleviating the diabetes-induced mechanical allodynia. Furthermore, we found that dorsal horn astrocytes express abundant N-myc downstream-regulated gene 2 (NDRG2), which contributes to astrocyte reactivity. NDRG2 was over-expressed in activated astrocytes in diabetic rats with mechanical allodynia. Intrathecal injection of RU486 prevented the over-expression of NDRG2, which reversed the astrocyte reactivity and diabetic tactile allodynia. These results suggest that glucocorticoid-mediated over-expression of NDRG2 may contribute to the activation of dorsal horn astrocytes, which play a crucial role in diabetic mechanical allodynia. Thus, inhibiting glucocorticoid receptors and/or astrocyte reactivity in the SDH may be a therapeutic strategy for treating diabetic tactile allodynia.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Glicerol, ACS reagent, ≥99.5%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glicerol, for molecular biology, ≥99.0%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glicerol, ReagentPlus®, ≥99.0% (GC)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Cloruro de sodio, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Acrilamida, suitable for electrophoresis, ≥99%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Cloruro de sodio, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
SAFC
Sodium chloride solution, 5 M
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glicerol solution, 83.5-89.5% (T)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glicerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Acrilamida, for molecular biology, ≥99% (HPLC)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glicerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glicerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
USP
Glicerol, United States Pharmacopeia (USP) Reference Standard
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Cloruro de sodio, 99.999% trace metals basis
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glicerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glicerol, FCC, FG
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Cloruro de sodio, BioUltra, for molecular biology, ≥99.5% (AT)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Acrilamida, suitable for electrophoresis, ≥99% (HPLC), powder
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glicerol, ≥99.5%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Cloruro de sodio, BioXtra, ≥99.5% (AT)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad