Merck
  • Immune augmentation and Dalton's Lymphoma tumor inhibition by glucans/glycans isolated from the mycelia and fruit body of Pleurotus ostreatus.
Immune augmentation and Dalton's Lymphoma tumor inhibition by glucans/glycans isolated from the mycelia and fruit body of Pleurotus ostreatus.
International immunopharmacology (2015-02-11)
K Sanjana P Devi, Birendra Behera, Debasish Mishra, Tapas K Maiti
RESUMEN
With the increase in cancer progression, alternatives in the medicinal field with minimal side effects need to be ascertained. In this context, for the first time novel glucans/glycans isolated from the mycelia and fruit body of Pleurotus ostreatus have been compared for their exquisite property as immunoceuticals. Glucans from both the sources displayed immunological functions which include lymphocyte proliferation, macrophage activation (nitric oxide production, ROS generation, phagocytosis, TNF-α production) as well as macrophage and NK cell mediated cytotoxicity. In vivo studies with Dalton's Lymphoma mice tumor model further enumerated the immune enhancing and tumor regression potential of the two glucan molecules. Highest tumor inhibition of about 75% and 71.4% were observed at 20mg/kg of mycelia and fruit body glucan/glycan treatments. A concomitant increase in the survival period of glucan treated tumor bearing mice was found to be primarily associated with immune boosting and apoptosis of cancerous cells. Both the glucan molecules exhibited similar degree of immune response at the systemic level with only subtle amount of differences in two dimensional in vitro cultures. Efficacy of glucans/glycans as immunomodulators may thereby provide decisive leads in strengthening the immune system along with other therapies.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Alcohol etílico puro 200, for molecular biology
Sigma-Aldrich
Alcohol etílico puro, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Alcohol etílico puro, 200 proof
Sigma-Aldrich
Alcohol etílico puro, 200 proof, meets USP testing specifications
Sigma-Aldrich
Alcohol etílico puro 190, for molecular biology
Sigma-Aldrich
Yoduro de propidio, ≥94.0% (HPLC)
Sigma-Aldrich
L-(−)-Fucosa, ≥98% (GC)
Sigma-Aldrich
Etanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Alcohol etílico puro, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Etanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Alcohol etílico puro, 200 proof, anhydrous, ≥99.5%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol solution, 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Etanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Etanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
USP
Etanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Alcohol etílico puro, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Supelco
Etanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Yoduro de propidio, ≥94% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)