Merck
Physiologically based pharmacokinetic modeling of impaired carboxylesterase-1 activity: effects on oseltamivir disposition.
Clinical pharmacokinetics (2014-08-12)
Zhe-Yi Hu, Andrea N Edginton, S Casey Laizure, Robert B Parker
RESUMEN
Human carboxylesterase-1 (CES1) is an enzyme that is primarily expressed in the liver, where it plays an important role in the metabolism of many commonly used medications. Ethanol (alcohol)-mediated inhibition of CES1 and loss-of-function polymorphisms in the CES1 gene can markedly reduce this enzyme's function. Such alterations in CES1 activity may have important effects on the disposition of substrate drugs. The aim of this study is to develop a physiologically based pharmacokinetic (PBPK) model to predict changes in CES1 substrate drug exposure in humans with CES1 activity impaired by ethanol or loss-of-function CES1 genetic polymorphisms. The antiviral drug oseltamivir, an ethyl ester prodrug that is rapidly converted in vivo to the active metabolite oseltamivir carboxylate (OSC) by CES1 was used as a probe drug for CES1 activity. Oseltamivir PBPK models integrating in vitro and in vivo data were developed and refined. Then the changes in oseltamivir and OSC exposure in humans with CES1 impaired by ethanol or polymorphisms were simulated using a PBPK model incorporating in vitro inhibition and enzyme kinetic data. Model assumptions were verified by comparison of simulations with observed and published data. A sensitivity analysis was performed to gain a mechanistic understanding of the exposure changes of oseltamivir and OSC. The simulated changes in oseltamivir and OSC exposures in humans with CES1 impaired by ethanol or polymorphism were similar to the observed data. The observed exposures to oseltamivir were increased by 46 and 37 % for the area under the plasma concentration-time curve from time zero to 6 h (AUC6) and from time zero to 24 h (AUC24), respectively, with co-administration of ethanol 0.6 g/kg. In contrast, only a slight change was observed in OSC exposure. The simulated data show the same trend as evidenced by greater change in exposures to oseltamivir (27 and 26 % for AUC(6) and AUC(24), [corrected] respectively) than OSC (≤6 %). The PBPK model of impaired CES1 activity correctly predicts observed human data. This model can be extended to predict the effects of drug interactions and other factors affecting the pharmacokinetics of other CES1 substrate drugs.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Alcohol etílico puro, for molecular biology
Sigma-Aldrich
Alcohol etílico puro, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Alcohol etílico puro, 200 proof
Sigma-Aldrich
Alcohol etílico puro, 200 proof, meets USP testing specifications
Sigma-Aldrich
Alcohol etílico puro, for molecular biology
Sigma-Aldrich
Etanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Alcohol etílico puro, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Etanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Alcohol etílico puro, 200 proof, anhydrous, ≥99.5%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol solution, 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Etanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Etanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
USP
Etanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Alcohol etílico puro, 190 proof, meets USP testing specifications
Supelco
Etanol, Pharmaceutical Secondary Standard; Certified Reference Material