Merck
Influence of chemical and structural features of low molecular weight heparins (LMWHs) on skin penetration.
International journal of pharmaceutics (2015-02-07)
S Franzè, C G M Gennari, P Minghetti, F Cilurzo
RESUMEN
Low molecular weight heparins (LMWHs) are obtained from unfractionated heparin (UFH) through different depolymerization methods (DM), which produce compounds having specific chemical features and biological activity. It is then supposed that LMWHs also exhibit different skin permeability properties. The current work aimed to get an insight on the in vitro passive diffusion through human epidermis of six commercially available LMWHs in comparison with UFH. The in vitro studies were performed using Franz diffusion cells. Heparins samples were assayed measuring the anti-factor Xa activity. Circular dichroism was used to evaluate the effect of the counter-ion (sodium or calcium) on the chain flexibility. The penetrated amounts after 24h (Q24) of sodium LMWHs were related to Mw by an exponential relationship (R=-0.758). The flux resulted dependent by DM following the rank order: β-elimination (8-11 mIU/cm(2)h range)>deaminative cleavage (5-7 mIU/cm(2)h range)>radical depolymerization (0.1mIU/cm(2)h). Finally, the calcium ion, reducing the chain flexibility, significantly affected the Q24 (0.001 ± 0.000 and 0.157 ± 0.049 IU/cm(2) for calcium and sodium nadroparin, respectively). Both the lower Mw and the introduction of new residues at the chain ends improved the skin penetration of LMWHs with respect to UFH (Q24=0.001 ± 0.001 IU/cm(2)), with bemiparin and enoxaparin being the most interesting compounds.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Phenol Red, ACS reagent
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad