Merck
Exome sequencing identifies SLCO2A1 mutations as a cause of primary hypertrophic osteoarthropathy.
American journal of human genetics (2011-12-27)
Zhenlin Zhang, Weibo Xia, Jinwei He, Zeng Zhang, Yaohua Ke, Hua Yue, Chun Wang, Hao Zhang, Jiemei Gu, Weiwei Hu, Wenzhen Fu, Yunqiu Hu, Miao Li, Yujuan Liu
RESUMEN
By using whole-exome sequencing, we identified a homozygous guanine-to-adenine transition at the invariant -1 position of the acceptor site of intron 1 (c.97-1G>A) in solute carrier organic anion transporter family member 2A1 (SLCO2A1), which encodes a prostaglandin transporter protein, as the causative mutation in a single individual with primary hypertrophic osteoarthropathy (PHO) from a consanguineous family. In two other affected individuals with PHO from two unrelated nonconsanguineous families, we identified two different compound heterozygous mutations by using Sanger sequencing. These findings confirm that SLCO2A1 mutations inactivate prostaglandin E(2) (PGE(2)) transport, and they indicate that mutations in SLCO2A1 are the pathogenic cause of PHO. Moreover, this study might also help to explain the cause of secondary hypertrophic osteoarthropathy.