Merck
The stem cell factor/Kit signalling pathway regulates mitochondrial function and energy expenditure.
Nature communications (2014-07-08)
Zan Huang, Hai-Bin Ruan, Li Xian, Weiqian Chen, Shujun Jiang, Anying Song, Qinghua Wang, Peiliang Shi, Xingxing Gu, Xiang Gao
RESUMEN
Cell growth is tightly coupled with mitochondrial biogenesis in order to maintain energy and organelle homeostasis. Receptor tyrosine kinase Kit and its ligand, stem cell factor (SCF), play a critical role in the growth and survival of multiple cell lineages. Here we report that the expression of SCF and Kit in adipose tissues is responsive to food availability and environmental temperature, and is altered in obese mice and human patients. Mice carrying a loss-of-function mutation in Kit develop obesity as a result of decreased energy expenditure. These phenotypes are associated with reduced PGC-1α expression and mitochondrial dysfunction in brown adipose tissue and skeletal muscle. We further demonstrate that SCF/Kit directly promotes Ppargc1a transcription and mitochondrial biogenesis. Blocking Kit signalling in mice decreases PGC-1α expression and thermogenesis, while overexpressing SCF systemically or specifically in brown adipose tissue increases thermogenesis and reduces weight gain. Collectively, these data provide mechanistic insight into the regulation of mitochondrial function by SCF/Kit signalling and lay a foundation for exploring SCF/Kit signalling as a therapeutic target for metabolic diseases.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sacarosa, for molecular biology, ≥99.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioXtra
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, BioUltra, for molecular biology, ≥99.5% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Anti-β-actina monoclonal antibody produced in mouse, clone AC-15, ascites fluid
En este momento no podemos mostrarle ni los precios ni la disponibilidad
USP
Sacarosa, United States Pharmacopeia (USP) Reference Standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Anti-α-tubulina monoclonal antibody produced in mouse, ascites fluid, clone B-5-1-2
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), Grade II, suitable for plant cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, Grade I, ≥99% (GC), suitable for plant cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, meets USP testing specifications
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Sacarosa, Pharmaceutical Secondary Standard; Certified Reference Material
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sacarosa, ACS reagent
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Millipore
Sacarosa, suitable for microbiology, ACS reagent, ≥99.0%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Sacarosa, puriss., meets analytical specification of Ph. Eur., BP, NF
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sacarosa, European Pharmacopoeia (EP) Reference Standard
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Supelco
Sacarosa, analytical standard, for enzymatic assay kit SCA20
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad