Merck
  • High-efficiency dye-sensitized solar cells: the influence of lithium ions on exciton dissociation, charge recombination, and surface states.
High-efficiency dye-sensitized solar cells: the influence of lithium ions on exciton dissociation, charge recombination, and surface states.
ACS nano (2010-10-07)
Qingjiang Yu, Yinghui Wang, Zhihui Yi, Ningning Zu, Jing Zhang, Min Zhang, Peng Wang
RESUMEN
We present a time-saving staining protocol based upon the low-toxicity and nonvolatile solvent dimethyl sulfoxide for a high-efficiency dye-sensitized solar cell, which is very critical for the roll-to-roll flexible cell production. Power conversion efficiencies of 11.7-12.1% were achieved under AM1.5G simulated sunlights. The intrinsic roles of lithium cations on efficiency enhancement were scrutinized by measuring transient absorption and electrical impedance spectroscopies. Our studies have revealed that lithium ions can enhance exciton dissociation at the energy-offset dye/titania interface effectively. Charge collection is not found to be a crucial current loss channel in our high-efficiency dye-sensitized solar cell.