- Syntheses of all the possible monomethyl ethers and several deoxyhalo analogues of methyl beta-lactoside as ligands for the Ricinus communis lectins.
Syntheses of all the possible monomethyl ethers and several deoxyhalo analogues of methyl beta-lactoside as ligands for the Ricinus communis lectins.
The synthesis of all the possible monomethyl ethers of methyl beta-lactoside (1) has been performed from 1 in a straightforward way, making use of the different reactivity of the hydroxyl groups in alkylation and stannylation reactions. In addition, the deoxyfluoro derivatives of 1 at positions, 6,3',4',epi-4', and 6' have been prepared by reaction of the appropriate substrates with diethylaminosulfur trifluoride or tetrabutylammonium fluoride. Finally, the 6-deoxyiodo and 6'-bromodeoxy analogues of 1 have also been prepared.