Merck
Doxepin concentrations in plasma and cerebrospinal fluid.
Journal of neural transmission (Vienna, Austria : 1996) (2011-02-26)
Robert Schomburg, Daniela Remane, Klaus Fassbender, Hans H Maurer, Jörg Spiegel
RESUMEN
Doxepin--like other antidepressant drugs (ADs)--shows a variable antidepressant effect in clinical practice. The cause for this variability is as yet unclear; however, pharmacokinetic factors such as the variable permeability of doxepin into the cerebrospinal fluid (CSF), may contribute to the difference in therapeutic efficacy. We measured and correlated the concentration of doxepin and its active metabolite nordoxepin in both the plasma and CSF. Plasma and CSF samples were taken simultaneously from 21 patients who were treated with doxepin due to different clinical indications. The plasma concentration of both doxepin and nordoxepin correlated significantly with the oral dosage of doxepin (doxepin: r = +0.66, p < 0.001; nordoxepin: r = +0.78, p < 0.0001; Spearman's correlation). Furthermore, we found significant correlations between the plasma and CSF concentrations of both doxepin (r = +0.71; p < 0.001; Pearson's correlation) and nordoxepin (r = +0.74; p < 0.001). These highly significant correlations between the plasma and CSF concentrations indicate a constant CSF permeability of doxepin and its active metabolite nordoxepin.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Nordoxepin hydrochloride, analytical standard, ≥98.0% (TLC), powder
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad