Merck
  • Quinine is a more potent inhibitor than quinidine in rat of the oxidative metabolic routes of methoxyphenamine which involve debrisoquine 4-hydroxylase.
Quinine is a more potent inhibitor than quinidine in rat of the oxidative metabolic routes of methoxyphenamine which involve debrisoquine 4-hydroxylase.
Xenobiotica; the fate of foreign compounds in biological systems (1991-11-01)
G Muralidharan, E M Hawes, G McKay, K K Midha
RESUMEN
1. Lewis rats (n = 7 or 8) were dosed with methoxyphenamine with and without prior administration of various doses of either quinine or its diastereomer quinidine. Methoxyphenamine and its N-desmethyl, O-desmethyl and aromatic 5-hydroxy metabolites were quantified in 0-24 h urine. 2. The oxidative routes of methoxyphenamine metabolism which had been previously shown to involve the debrisoquine/sparteine isoenzyme, namely O-demethylation and 5-hydroxylation, were both significantly inhibited by quinine. The inhibition was selective in that N-demethylation which does not involve this isoenzyme was not affected by quinine. 3. Quinidine which had been previously shown at a relatively high dose (80 mg/kg) to affect the three metabolic routes of methoxyphenamine in a similar fashion was ineffective in this regard at a 25 mg/kg dose. Quinine more effectively inhibited the O-demethylation and 5-hydroxylation of methoxyphenamine than did quinidine, and its inhibition was marked at the lowest dose examined, 12.5 mg/kg. 4. As quinidine is a more potent inhibitor than quinine of debrisoquine 4-hydroxylase in man, the rat should be used only with full realization of its limitations when investigating substrates metabolized by this isoenzyme.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methoxyphenamine hydrochloride, ≥99%