Merck
  • Topical calcipotriene 0.005% and betamethasone dipropionate 0.064% maintains efficacy of etanercept after step-down dose in patients with moderate-to-severe plaque psoriasis: results of an open label trial.
Topical calcipotriene 0.005% and betamethasone dipropionate 0.064% maintains efficacy of etanercept after step-down dose in patients with moderate-to-severe plaque psoriasis: results of an open label trial.
Journal of drugs in dermatology : JDD (2011-08-06)
Leon H Kircik
RESUMEN
Etanercept is approved for the treatment of plaque psoriasis at a subcutaneous (SC) dosage of 50 mg twice-weekly for three months, followed by 50 mg SC once-weekly thereafter. It is of note, however, that many patients experience loss of efficacy when they step down to etanercept 50 mg/week, often instigating a switch to another biological. The current pilot study investigated the adjunctive use of a topical calcipotriene 0.005% and betamethasone dipropionate 0.064% combination ointment, approved for the treatment of psoriasis, to sustain the original efficacy of etanercept by augmenting response to the 50 mg/week SC dose, thus stabilizing the disease. In this single-center, open-label study, subjects (n=20) underwent 12 weeks treatment with etanercept 100 mg/week (50 mg, 2x week; weeks -12 to -1), followed by etanercept 50 mg/week maintenance therapy for 40 weeks (weeks 0 to 40). Subjects were followed at four-week intervals. Starting at week 4, subjects who demonstrated an increase from baseline (week 0) body surface area (BSA) of is greater than 2% initiated therapy with calcipotriene 0.005% and betamethasone dipropionate 0.064% ointment for four weeks. The study is limited by its small sample size, open-label nature, and lack of blinding. Mean BSA involvement decreased significantly from week -12 to 0 with etanercept 50 mg twice a week. At week 4, on the etanercept 50 mg/week dose, mean BSA increased to 9.42 ± 9.39 compared to week 0. With introduction of calcipotriene 0.005%/betamethasone dipropionate 0.064% ointment at week 4, mean BSA decreased to 4.62 ± 8.19 by week 24 and was relatively stable for the remainder of the study period. Similarly, mean PASI (Psoriasis Area and Severity Index) scores improved from week -12 to week 0, increased at week 4, then decreased significantly by week 24 with adjunctive topical treatment. Topical calcipotriene 0.005% and betamethasone dipropionate 0.064% ointment is a safe and effective adjunct to etanercept 50 mg/week maintenance therapy.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Betamethasone 17,21-dipropionate
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Betamethasone dipropionate, European Pharmacopoeia (EP) Reference Standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad